1-Benzhydrylazetidin-3-one 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: 1-Benzhydryl-3-azetidinone, 1-Diphenylmethyl-3-azetidinone

OR9090-5G OR9090-1G OR9090-10G
APOSOR9090-5GEA 20 GBP
APOSOR9090-5G APOSOR9090-1G APOSOR9090-10G
1-Benzhydrylazetidin-3-one 98%
1-Benzhydrylazetidin-3-one
Formula: C₁₆H₁₅NO
MW: 237.30 g/mol
Boiling Pt: 350…355 °C
Density: 1.182 g/cm³ (25 °C)
Flash Pt: 156 °C
Storage Temperature: Ambient
MDL Number: MFCD00964539
CAS Number: 40320-60-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR