tert-Butyl-2-methyl-4-oxopiperidine-1-carboxylate 97%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: 2-Methylpiperidin-4-one, N-BOC protected, tert-Butyl 2-methyl-4-oxopiperidine-1-carboxylate

OR9058-25G
APOSOR9058-25GEA 99 GBP
APOSOR9058-25G
tert-Butyl-2-methyl-4-oxopiperidine-1-carboxylate 97%
tert-Butyl-2-methyl-4-oxopiperidine-1-carboxylate
Formula: C₁₁H₁₉NO₃
MW: 213.2735 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04035608
CAS Number: 190906-92-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR