1-(3-Chlorophenyl)-1H-pyrrole 95%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR905325-1G
APOSOR905325-1GEA 252 GBP
APOSOR905325-1G
1-(3-Chlorophenyl)-1H-pyrrole 95%
1-(3-Chlorophenyl)-1H-pyrrole
Formula: C₁₀H₈ClN
MW: 177.63 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02665247
CAS Number: 41910-45-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR