2-Amino-3-(dimethylamino)pyrazine 95%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR905054-1G OR905054-250MG
APOSOR905054-1GEA 336 GBP
APOSOR905054-1G APOSOR905054-250MG
2-Amino-3-(dimethylamino)pyrazine 95%
2-Amino-3-(dimethylamino)pyrazine
Formula: C₆H₁₀N₄
Storage Temperature: Ambient
CAS Number: 89488-74-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR