Imidazo[1,5-a]pyridine-6-carboxylic acid 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR903526-1G OR903526-250MG
APOSOR903526-1GEA 328 GBP
APOSOR903526-1G APOSOR903526-250MG
Imidazo[1,5-a]pyridine-6-carboxylic acid 98%
Imidazo[1,5-a]pyridine-6-carboxylic acid
Formula: C₈H₆N₂O₂
MW: 162.15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11042992
CAS Number: 256935-76-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR