3-Amino-2-hydroxybenzonitrile 95%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR903180-1G
APOSOR903180-1GEA 649 GBP
APOSOR903180-1G
3-Amino-2-hydroxybenzonitrile 95%
3-Amino-2-hydroxybenzonitrile
Formula: C₇H₆N₂O
MW: 134.135 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD02261942
CAS Number: 67608-57-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR