6-Phenyl-2-pyridone 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: 6-Phenylpyridin-2(1H)-one

OR901687-1G OR901687-5G
APOSOR901687-1GEA 158 GBP
APOSOR901687-1G APOSOR901687-5G
6-Phenyl-2-pyridone 98%
6-Phenyl-2-pyridone
Formula: C₁₁H₉NO
MW: 171.20 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00059760
CAS Number: 19006-82-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR