4-Methoxynaphthalen-1-amine 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR900640-1G
APOSOR900640-1GEA 263 GBP
APOSOR900640-1G
4-Methoxynaphthalen-1-amine 98%
4-Methoxynaphthalen-1-amine
Formula: C₁₁H₁₁NO
MW: 173.211 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09743196
CAS Number: 16430-99-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR