2-Cyanobenzenesulphonyl chloride

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

OR8940-25G
APOSOR8940-25GEA 109 GBP
APOSOR8940-25G
2-Cyanobenzenesulphonyl chloride
2-Cyanobenzenesulphonyl chloride
Formula: C₇H₄ClNO₂S
Melting Pt: 65…69 °C
Storage Temperature: Ambient
MDL Number: MFCD00052341
CAS Number: 69360-26-5
UN: 3261
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR