2-(2-(Piperazin-1-yl)ethoxy)ethanol

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: 1-[2-(2-Hydroxyethoxy)ethyl]piperazine, 1-Hydroxyethylethoxypiperazine

OR8790-100G OR8790-25G OR8790-500G
APOSOR8790-100GEA 52.5 GBP
APOSOR8790-100G APOSOR8790-25G APOSOR8790-500G
2-(2-(Piperazin-1-yl)ethoxy)ethanol
2-(2-(Piperazin-1-yl)ethoxy)ethanol
Formula: C₈H₁₈N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD00074931
CAS Number: 13349-82-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR