2-Amino-5,6,7,8-tetrahydroquinazolin-4-ol

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: 2-Amino-4-hydroxy-5,6,7,8-tetrahydroquinazoline

OR8486-100MG
APOSOR8486-100MGEA 16.8 GBP
APOSOR8486-100MG
2-Amino-5,6,7,8-tetrahydroquinazolin-4-ol
2-Amino-5,6,7,8-tetrahydroquinazolin-4-ol
Formula: C₈H₁₁N₃O
MW: 165.2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00665850
CAS Number: 33081-07-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR