3-Phenoxybenzyl chloride 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

OR8418-25G OR8418-5G OR8418-1G
APOSOR8418-25GEA 195 GBP
APOSOR8418-25G APOSOR8418-5G APOSOR8418-1G
3-Phenoxybenzyl chloride 98%
3-Phenoxybenzyl chloride
Formula: C₁₃H₁₁ClO
Boiling Pt: 141…142 °C/5mmHg
Density: 1.189 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00040866
CAS Number: 53874-66-1
EINECS: 258-831-1
UN: 1760
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR