6-Nitro-m-cresol

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: 5-Methyl-2-nitrophenol, 1-Hydroxy-3-methyl-6-nitrobenzene

OR8232-100G
APOSOR8232-100GEA 46.2 GBP
APOSOR8232-100G
6-Nitro-m-cresol
6-Nitro-m-cresol
Formula: C₇H₇NO₃
MW: 153.14 g/mol
Melting Pt: 53…56 °C
Flash Pt: 109 °C
Storage Temperature: Ambient
MDL Number: MFCD00007111
CAS Number: 700-38-9
EINECS: 211-843-0
UN: 2446
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR