2-Methyl-5-[(pyrrolidin-1-yl)sulphonyl]-3-furoic acid 95%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

OR8134-100MG
APOSOR8134-100MGEA 47.3 GBP
APOSOR8134-100MG
2-Methyl-5-[(pyrrolidin-1-yl)sulphonyl]-3-furoic acid 95%
2-Methyl-5-[(pyrrolidin-1-yl)sulphonyl]-3-furoic acid
Formula: C₁₀H₁₃NO₅S
Melting Pt: 185 °C
Storage Temperature: Ambient
MDL Number: MFCD02180785
CAS Number: 306936-43-6
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR