Ethyl 1-(3-aminobenzyl)piperidine-4-carboxylate 97%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: 1-(3-Aminobenzyl)-4-(ethoxycarbonyl)piperidine, 3-{[4-Ethoxycarbonyl)piperidin-1-yl]methyl}aniline

OR8090-1G OR8090-250MG OR8090-5G
APOSOR8090-1GEA 73.5 GBP
APOSOR8090-1G APOSOR8090-250MG APOSOR8090-5G
Ethyl 1-(3-aminobenzyl)piperidine-4-carboxylate 97%
Ethyl 1-(3-aminobenzyl)piperidine-4-carboxylate
Formula: C₁₅H₂₂N₂O₂
MW: 262.347 g/mol
Melting Pt: 49…53 °C
Storage Temperature: Ambient
MDL Number: MFCD02180894
CAS Number: 306937-22-4
UN: 3259
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR