6-Amino-3-bromo-2-methylpyridine

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

OR7864-100G OR7864-25G OR7864-500G
APOSOR7864-100GEA 137 GBP
APOSOR7864-100G APOSOR7864-25G APOSOR7864-500G
6-Amino-3-bromo-2-methylpyridine
6-Amino-3-bromo-2-methylpyridine
Formula: C₆H₇BrN₂
Melting Pt: 74…79 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00068230
CAS Number: 42753-71-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR