2-Hydroxyisonicotinonitrile

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: 4-Cyano-2-hydroxypyridine, 4-Cyano-2(1H)-pyridinone, 2-Hydroxypyridine-4-carbonitrile

OR7613-250MG
APOSOR7613-250MGEA 15.8 GBP
APOSOR7613-250MG
2-Hydroxyisonicotinonitrile
2-Hydroxyisonicotinonitrile
Formula: C₆H₄N₂O
MW: 120.109 g/mol
Melting Pt: 285…288 °C
Storage Temperature: Ambient
MDL Number: MFCD04037920
CAS Number: 95891-29-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR