Methyl 4-(3-hydroxyprop-1-yn-1-yl)benzoate

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

OR7570-250MG OR7570-1G
APOSOR7570-250MGEA 23.1 GBP
APOSOR7570-250MG APOSOR7570-1G
Methyl 4-(3-hydroxyprop-1-yn-1-yl)benzoate
Methyl 4-(3-hydroxyprop-1-yn-1-yl)benzoate
Formula: C₁₁H₁₀O₃
Storage Temperature: Ambient
MDL Number: MFCD00168848
CAS Number: 61266-36-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR