1-Bromononane 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: 1-Nonyl bromide

OR7540-25G OR7540-100G OR7540-500G
APOSOR7540-25GEA 65.1 GBP
APOSOR7540-25G APOSOR7540-100G APOSOR7540-500G
1-Bromononane 98%
1-Bromononane
Formula: BrH₂C(CH₂)₇CH₃
Boiling Pt: 200…201 °C
Density: 1.084 g/cm³ (25 °C)
Flash Pt: 90 °C
Storage Temperature: Ambient
MDL Number: MFCD00000278
CAS Number: 693-58-3
EINECS: 211-755-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR