5-Formyl-2-nitrobenzoic acid

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

OR72161-1G
APOSOR72161-1GEA 567 GBP
APOSOR72161-1G
5-Formyl-2-nitrobenzoic acid
5-Formyl-2-nitrobenzoic acid
Formula: C₈H₅NO₅
MW: 195.13 g/mol
MDL Number: MFCD26855792
CAS Number: 1289007-91-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR