4,4'-Bis(dimethylamino)benzophenone

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: 4,4′-Bis(dimethylamino)benzophenone, 4,4-Bis(dimethylamino)benzophenone, 4,4'-Di(dimethylamino)benzophenone, Michler's ketone, Bis[4-(dimethylamino)phenyl]methanone

OR72059-100G OR72059-500G
APOSOR72059-100GEA 60.9 GBP
APOSOR72059-100G APOSOR72059-500G
4,4'-Bis(dimethylamino)benzophenone
4,4'-Bis(dimethylamino)benzophenone
Formula: C₁₇H₂₀N₂O
Boiling Pt: 360 °C
Melting Pt: 171 - 176 °C
Flash Pt: 220 °C
MDL Number: MFCD00008312
CAS Number: 90-94-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR