1-Cyclopropylethylamine hydrochloride

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

OR7051-10G OR7051-1G OR7051-5G
APOSOR7051-10GEA 416 GBP
APOSOR7051-10G APOSOR7051-1G APOSOR7051-5G
1-Cyclopropylethylamine hydrochloride
1-Cyclopropylethylamine hydrochloride
Formula: C₅H₁₂ClN
Storage Temperature: Ambient
MDL Number: MFCD06685893
CAS Number: 42390-64-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR