1-(2-Aminoethyl)-1H-benzimidazole hydrochloride

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: 2-(1H-Benzimidazol-1-yl)ethylamine hydrochloride

OR7037-250MG
APOSOR7037-250MGEA 29.4 GBP
APOSOR7037-250MG
1-(2-Aminoethyl)-1H-benzimidazole hydrochloride
1-(2-Aminoethyl)-1H-benzimidazole hydrochloride
Formula: C₉H₁₁N₃·ClH
Storage Temperature: Ambient
MDL Number: MFCD07366475
CAS Number: 1085300-74-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR