3,4-Dichlorobenzophenone

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: (3,4-Dichlorophenyl)(phenyl)methanone

OR70083-100G OR70083-25G
APOSOR70083-100GEA 63 GBP
APOSOR70083-100G APOSOR70083-25G
3,4-Dichlorobenzophenone
3,4-Dichlorobenzophenone
Formula: C₁₃H₈Cl₂O
Melting Pt: 100…104 °C
Storage Temperature: Ambient
MDL Number: MFCD00000552
CAS Number: 6284-79-3
EINECS: 228-509-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR