3-Bromo-5-methoxybenzyl bromide

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

OR70061-1G OR70061-5G
APOSOR70061-1GEA 42 GBP
APOSOR70061-1G APOSOR70061-5G
3-Bromo-5-methoxybenzyl bromide
3-Bromo-5-methoxybenzyl bromide
Formula: C₈H₈Br₂O
Storage Temperature: Refrigerator
MDL Number: MFCD12761608
CAS Number: 59297-29-9
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR