1-Cycloheptylpiperazine 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

OR6905-1G
APOSOR6905-1GEA 31.5 GBP
APOSOR6905-1G
1-Cycloheptylpiperazine 98%
1-Cycloheptylpiperazine
Formula: C₁₁H₂₂N₂
Boiling Pt: 102 °C/2.5mmHg
Storage Temperature: Ambient
MDL Number: MFCD01076573
CAS Number: 21043-42-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR