5-Amino-3-(fur-2-yl)isoxazole

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: 3-(Furan-2-yl)isoxazol-5-amine

OR6697-150MG OR6697-50MG
APOSOR6697-150MGEA 106 GBP
APOSOR6697-150MG APOSOR6697-50MG
5-Amino-3-(fur-2-yl)isoxazole
5-Amino-3-(fur-2-yl)isoxazole
Formula: C₇H₆N₂O₂
Melting Pt: 99 °C
Storage Temperature: Ambient
MDL Number: MFCD01754641
CAS Number: 33866-44-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR