4-Bromo-3-iodobenzaldehyde

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

OR63029-250MG OR63029-1G OR63029-5G
APOSOR63029-250MGEA 42 GBP
APOSOR63029-250MG APOSOR63029-1G APOSOR63029-5G
4-Bromo-3-iodobenzaldehyde
4-Bromo-3-iodobenzaldehyde
Formula: C₇H₄BrIO
Melting Pt: 127 °C
Storage Temperature: Ambient
MDL Number: MFCD18391620
CAS Number: 873387-81-6
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR