3-Nitrophthalic anhydride 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

OR63021-100G OR63021-250G OR63021-25G
APOSOR63021-100GEA 31.5 GBP
APOSOR63021-100G APOSOR63021-250G APOSOR63021-25G
3-Nitrophthalic anhydride 98%
3-Nitrophthalic anhydride
Formula: C₈H₃NO₅
Melting Pt: 163…165 °C
Storage Temperature: Ambient
MDL Number: MFCD00005921
CAS Number: 641-70-3
EINECS: 211-373-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR