2-(Pyridin-3-yl)acetic acid hydrochloride 95%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: Pyridyl-3-acetic acid hydrochloride, 3-Pyridylacetic acid hydrochloride

OR6262-25G OR6262-100G
APOSOR6262-25GEA 15.8 GBP
APOSOR6262-25G APOSOR6262-100G
2-(Pyridin-3-yl)acetic acid hydrochloride 95%
2-(Pyridin-3-yl)acetic acid hydrochloride
Formula: C₇H₈ClNO₂
Melting Pt: 161…163 °C
Storage Temperature: Ambient
MDL Number: MFCD00012819
CAS Number: 6419-36-9
EINECS: 229-148-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR