4-Aminopyridine-2-carboxylic acid 97%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: 4-Aminopyridine-2-carboxylic Acid

OR6260-1G OR6260-250MG
APOSOR6260-1GEA 56.7 GBP
APOSOR6260-1G APOSOR6260-250MG
4-Aminopyridine-2-carboxylic acid 97%
4-Aminopyridine-2-carboxylic acid
Formula: C₆H₆N₂O₂
Melting Pt: 319 °C
Storage Temperature: Ambient
MDL Number: MFCD02685671
CAS Number: 100047-36-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR