Tributyrin (glycerol tributyrate) ≥97%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Tributyrin, Glyceryl tributyrate, Glycerol tributyrate, Propane-1,2,3-triyl tributyrate

OR62132-25G OR62132-100G OR62132-500G
APOSOR62132-25GEA 23.1 GBP
APOSOR62132-25G APOSOR62132-100G APOSOR62132-500G
Tributyrin (glycerol tributyrate) ≥97%
Tributyrin (Glycerol tributyrate)
Formula: C₁₅H₂₆O₆
Boiling Pt: 129-131 °C/0.03mm
Melting Pt: -75 °C
Density: 1.032
Flash Pt: 174 °C
MDL Number: MFCD00009392
CAS Number: 60-01-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR