4-Iodobenzylamine 95+%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: (4-Iodophenyl)methanamine

OR6197-1G
APOSOR6197-1GEA 50.5 GBP
APOSOR6197-1G
4-Iodobenzylamine 95+%
4-Iodobenzylamine
Formula: C₇H₈IN
MW: 233.05 g/mol
Boiling Pt: 113…115 °C/4mmHg
Melting Pt: 46…48 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00047933
CAS Number: 39959-59-6
UN: 3259
ADR: 8 II

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR