3-Methyl-2-nitrobenzoic acid

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

OR61446-25G
APOSOR61446-25GEA 15.8 GBP
APOSOR61446-25G
3-Methyl-2-nitrobenzoic acid
2-Nitro-m-toluic acid
Formula: C₈H₇NO₄
Melting Pt: 220…223 °C
Storage Temperature: Ambient
MDL Number: MFCD00007180
CAS Number: 5437-38-7
EINECS: 226-610-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR