(±)-3-(Boc-amino)piperidine 97%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: tert-Butyl piperidin-3-ylcarbamate, 3-Aminopiperidine, 3-BOC protected

OR6133-1G OR6133-25G OR6133-5G
APOSOR6133-1GEA 15.8 GBP
APOSOR6133-1G APOSOR6133-25G APOSOR6133-5G
(±)-3-(Boc-amino)piperidine 97%
(±)-3-(Boc-amino)piperidine
Formula: C₁₀H₂₀N₂O₂
Melting Pt: 102…108 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03839941
CAS Number: 172603-05-3
UN: 3077
ADR: 9+EHS,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR