Dodecylamine 97%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

OR61300-100G OR61300-1KG OR61300-500G
APOSOR61300-100GEA 57.8 GBP
APOSOR61300-100G APOSOR61300-1KG APOSOR61300-500G
Dodecylamine 97%
Dodecylamine
Formula: H₃C(CH₂)₁₀CH₂NH₂
Boiling Pt: 247…249 °C
Melting Pt: 27…31 °C
Density: 0.806 g/cm³ (25 °C)
Flash Pt: 115 °C
Storage Temperature: Ambient
MDL Number: MFCD00008154
CAS Number: 124-22-1
EINECS: 204-690-6
UN: 3259
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR