2-methyl-3,4-dihydro-2H-1,4-benzothiazine

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR61156-1G
APOSOR61156-1GEA 247 GBP
APOSOR61156-1G
2-methyl-3,4-dihydro-2H-1,4-benzothiazine
2-methyl-3,4-dihydro-2H-1,4-benzothiazine
Formula: C₉H₁₁NS
Boiling Pt: 77 °C/0.01mmHg
Density: 1.084 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD12027245
CAS Number: 58960-00-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR