2,5-Dimethylthiazole

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

OR61055-5G
APOSOR61055-5GEA 46.2 GBP
APOSOR61055-5G
2,5-Dimethylthiazole
2,5-Dimethylthiazole
Formula: C₅H₇NS
Boiling Pt: 148…153 °C
Density: 1.057 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00130120
CAS Number: 4175-66-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR