2-Amino-4-methoxynicotinonitrile

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

OR6055-1G OR6055-5G
APOSOR6055-1GEA 85.6 GBP
APOSOR6055-1G APOSOR6055-5G
2-Amino-4-methoxynicotinonitrile
2-Amino-4-methoxynicotinonitrile
Formula: C₇H₇N₃O
Melting Pt: 211 °C
Storage Temperature: Ambient
MDL Number: MFCD00975441
CAS Number: 98651-70-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR