5-Bromo-1H-pyrazolo[3,4-c]pyridine

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

OR60159-5G OR60159-1G
APOSOR60159-5GEA 47.3 GBP
APOSOR60159-5G APOSOR60159-1G
5-Bromo-1H-pyrazolo[3,4-c]pyridine
5-Bromo-1H-pyrazolo[3,4-c]pyridine
Formula: C₆H₄BrN₃
Storage Temperature: Refrigerator
MDL Number: MFCD11518975
CAS Number: 929617-35-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR