2-(Aminomethyl)-3-bromo-5-methylpyridine hydrochloride

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: (3-Bromo-5-methylpyridin-2-yl)methylamine hydrochloride

OR60142-100MG
APOSOR60142-100MGEA 168 GBP
APOSOR60142-100MG
2-(Aminomethyl)-3-bromo-5-methylpyridine hydrochloride
2-(Aminomethyl)-3-bromo-5-methylpyridine hydrochloride
Formula: C₇H₉BrN₂·ClH
Storage Temperature: Refrigerator
MDL Number: MFCD17214207
CAS Number: 1257535-62-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR