N-Boc-L-valine 97%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Boc-Val-OH, BOC-L-Valine

OR60097-500G
APOSOR60097-500GEA 64.1 GBP
APOSOR60097-500G
N-Boc-L-valine 97%
N-Boc-L-valine
Formula: C₁₀H₁₉NO₄
MW: 217.3 g/mol
Melting Pt: 78…80 °C
Storage Temperature: Ambient
MDL Number: MFCD00065605
CAS Number: 13734-41-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR