4-Bromo-1-methyl-1H-indazole 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

OR60015-1G
APOSOR60015-1GEA 23.1 GBP
APOSOR60015-1G
4-Bromo-1-methyl-1H-indazole 98%
4-Bromo-1-methyl-1H-indazole
Formula: C₈H₇BrN₂
MW: 211.0586 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09870042
CAS Number: 365427-30-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR