2-(Hydroxymethyl)-1,3-oxazole

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: 2-Oxazolemethanol

OR59845-1G OR59845-250MG
APOSOR59845-1GEA 183 GBP
APOSOR59845-1G APOSOR59845-250MG
2-(Hydroxymethyl)-1,3-oxazole
2-(Hydroxymethyl)-1,3-oxazole
Formula: C₄H₅NO₂
Storage Temperature: Ambient
MDL Number: MFCD08703642
CAS Number: 130551-92-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR