1-Bromo-1-cycloheptene

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR59406-1G
APOSOR59406-1GEA 132 GBP
APOSOR59406-1G
1-Bromo-1-cycloheptene
1-Bromo-1-cycloheptene
Formula: C₇H₁₁Br
Boiling Pt: 81…82 °C/2mmHg
Storage Temperature: Refrigerator
MDL Number: MFCD09878419
CAS Number: 18317-64-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR