4-Bromo-N-ethylbenzenesulphonamide 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR5692-5G
APOSOR5692-5GEA 82.9 GBP
APOSOR5692-5G
4-Bromo-N-ethylbenzenesulphonamide 98%
4-Bromo-N-ethylbenzenesulphonamide
Formula: C₈H₁₀BrNO₂S
MW: 264.1395 g/mol
Melting Pt: 69…71 °C
Storage Temperature: Ambient
MDL Number: MFCD01215100
CAS Number: 1984-25-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR