2-Acetamidoisonicotinic acid 97%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

OR5582-1G OR5582-250MG
APOSOR5582-1GEA 21.1 GBP
APOSOR5582-1G APOSOR5582-250MG
2-Acetamidoisonicotinic acid 97%
2-Acetamidoisonicotinic acid
Formula: C₈H₈N₂O₃
Melting Pt: 286…290 °C
Storage Temperature: Ambient
MDL Number: MFCD00233713
CAS Number: 54221-95-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR