2-Bromophenol 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

OR5397-100G OR5397-25G OR5397-500G
APOSOR5397-100GEA 42 GBP
APOSOR5397-100G APOSOR5397-25G APOSOR5397-500G
2-Bromophenol 98%
2-Bromophenol
Formula: C₆H₅BrO
Boiling Pt: 194…195 °C
Melting Pt: 44380 °C
Density: 1.49 g/cm³ (25 °C)
Flash Pt: 42 °C
Storage Temperature: Ambient
MDL Number: MFCD00002146
CAS Number: 95-56-7
EINECS: 202-432-7
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR