1H-Imidazol-1-amine hydrochloride 97%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR52986-250MG
APOSOR52986-250MGEA 101 GBP
APOSOR52986-250MG
1H-Imidazol-1-amine hydrochloride 97%
1H-Imidazol-1-amine hydrochloride
Formula: C₃H₅N₃·ClH
Storage Temperature: Ambient
MDL Number: MFCD26390998
CAS Number: 83279-44-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR