4-Bromophenylhydrazinium chloride 95%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: (4-Bromophenyl)hydrazine hydrochloride, 4-Bromophenylhydrazine hydrochloride, 1-Bromo-4-hydrazinobenzene hydrochloride, 1-(4-Bromophenyl)hydrazine hydrochloride

OR5273-100G OR5273-500G
APOSOR5273-100GEA 55.7 GBP
APOSOR5273-100G APOSOR5273-500G
4-Bromophenylhydrazinium chloride 95%
4-Bromophenylhydrazinium chloride
Formula: C₆H₈BrClN₂
Melting Pt: 211…214 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012941
CAS Number: 622-88-8
UN: 3261
ADR: 8,II

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR